Assay Details
| Gene Symbol: PMEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319557.1 | NP_001306486.1 | 3 - 4 | 615 | 67 | |
| XM_015431013.1 | XP_015286499.1 | 2 - 3 | 879 | 67 | ||
| XM_015431014.1 | XP_015286500.1 | 2 - 3 | 849 | 67 | ||
| GenBank mRNA | AB168730.1 | - | 3 - 4 | 615 | 67 | |
More Information
Set Membership: |
|

PMEL
Genomic Map