Assay Details
| Gene Symbol: SHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541642.2 | XP_005541699.1 | 9 - 10 | 2066 | 82 | |
| XM_015456546.1 | XP_015312032.1 | 8 - 9 | 2015 | 82 | ||
| XM_015456551.1 | XP_015312037.1 | 10 - 11 | 1154 | 82 | ||
| XM_015456562.1 | XP_015312048.1 | 9 - 10 | 1100 | 82 | ||
More Information
Set Membership: |
|

SHC1
Genomic Map