Assay Details
| Gene Symbol: PXYLP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005545944.1 | XP_005546001.1 | 4 - 5 | 560 | 71 | |
| XM_005545945.2 | XP_005546002.1 | 4 - 5 | 1794 | 71 | ||
More Information
Set Membership: |
|

PXYLP1
Genomic Map