Assay Details
| Gene Symbol: NT5C3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549835.2 | XP_005549892.1 | 6 - 7 | 710 | 133 | |
| XM_005549836.2 | XP_005549893.1 | 8 - 9 | 1323 | 133 | ||
| XM_005549837.2 | XP_005549894.1 | 7 - 8 | 1133 | 133 | ||
| XM_005549838.1 | XP_005549895.1 | 7 - 8 | 674 | 133 | ||
| GenBank mRNA | AB173701.1 | - | 7 - 8 | 653 | 133 | |
More Information
Set Membership: |
|

NT5C3A
Genomic Map