Assay Details
| Gene Symbol: PTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005550873.2 | XP_005550930.1 | 2 - 3 | 125 | 76 | |
| XM_005550874.2 | XP_005550931.1 | 2 - 3 | 125 | 76 | ||
| XM_005550875.2 | XP_005550932.1 | 2 - 3 | 524 | 76 | ||
| XM_005550876.2 | XP_005550933.1 | 2 - 3 | 524 | 76 | ||
More Information
Set Membership: |
|

PTN
Genomic Map