Assay Details
| Gene Symbol: TNFRSF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544760.2 | XP_005544817.1 | 2 - 3 | 328 | 85 | |
| XM_005544761.2 | XP_005544818.1 | 2 - 3 | 328 | 85 | ||
More Information
Set Membership: |
|

TNFRSF1B
Genomic Map