Assay Details
| Gene Symbol: HPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578799.2 | XP_005578856.1 | 7 - 8 | 937 | 69 | |
| XM_005578800.2 | XP_005578857.1 | 7 - 8 | 874 | 69 | ||
More Information
Set Membership: |
|

HPX
Genomic Map