Assay Details
| Gene Symbol: SIPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577253.2 | XP_005577310.1 | 12 - 13 | 2932 | 73 | |
| XM_005577254.2 | XP_005577311.1 | 12 - 13 | 3488 | 73 | ||
| XM_005577255.2 | XP_005577312.1 | 12 - 13 | 3061 | 73 | ||
More Information
Set Membership: |
|

SIPA1
Genomic Map