Assay Details
| Gene Symbol: TAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544220.2 | XP_005544277.1 | 3 - 4 | 441 | 93 | |
| XM_005544221.2 | XP_005544278.1 | 2 - 3 | 416 | 93 | ||
| XM_005544222.2 | XP_005544279.1 | 2 - 3 | 424 | 93 | ||
More Information
Set Membership: |
|

TAF12
Genomic Map