Assay Details
| Gene Symbol: GSK3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548051.2 | XP_005548108.1 | 1 - 2 | 604 | 64 | |
| XM_005548052.2 | XP_005548109.1 | 1 - 2 | 604 | 64 | ||
More Information
Set Membership: |
|

GSK3B
Genomic Map