Assay Details
| Gene Symbol: WARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283107.1 | NP_001270036.1 | 7 - 8 | 951 | 90 | |
| XM_005562211.1 | XP_005562268.1 | 7 - 8 | 1017 | 90 | ||
| XM_005562212.2 | XP_005562269.1 | 7 - 8 | 1210 | 90 | ||
| XM_015453230.1 | XP_015308716.1 | 8 - 9 | 1768 | 90 | ||
| XM_015453231.1 | XP_015308717.1 | 7 - 8 | 1019 | 90 | ||
| XM_015453232.1 | XP_015308718.1 | 7 - 8 | 919 | 90 | ||
| GenBank mRNA | AB169365.1 | - | 7 - 8 | 951 | 90 | |
| AB170766.1 | - | 7 - 8 | 945 | 90 | ||
More Information
Set Membership: |
|

WARS1
Genomic Map