Assay Details
| Gene Symbol: IDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574098.2 | XP_005574155.1 | 9 - 10 | 1452 | 109 | |
| XM_005574099.2 | XP_005574156.1 | 9 - 10 | 1522 | 109 | ||
More Information
Set Membership: |
|

IDH1
Genomic Map