Assay Details
| Gene Symbol: CSNK2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553513.2 | XP_005553570.1 | 6 - 7 | 847 | 60 | |
| XM_005553514.1 | XP_005553571.1 | 6 - 7 | 890 | 60 | ||
More Information
Set Membership: |
|

CSNK2B
Genomic Map