Assay Details
| Gene Symbol: HEY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551729.2 | XP_005551786.1 | 4 - 5 | 476 | 95 | |
| XM_005551730.2 | XP_005551787.1 | 4 - 5 | 638 | 95 | ||
| XM_005551731.2 | XP_005551788.1 | 4 - 5 | 698 | 95 | ||
More Information
Set Membership: |
|

HEY2
Genomic Map