Assay Details
| Gene Symbol: NDUFA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569865.2 | XP_005569922.1 | 3 - 4 | 391 | 64 | |
| XM_005569866.2 | XP_005569923.1 | 3 - 4 | 391 | 64 | ||
More Information
Set Membership: |
|

NDUFA9
Genomic Map