Assay Details
| Gene Symbol: PRPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319562.1 | NP_001306491.1 | 1 - 2 | 237 | 60 | |
| XM_005592966.2 | XP_005593023.2 | 1 - 2 | 261 | 60 | ||
| GenBank mRNA | AB169827.1 | - | 1 - 2 | 237 | 60 | |
More Information
Set Membership: |
|

PRPS2
Genomic Map