Assay Details
| Gene Symbol: CDKN1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015434292.1 | XP_015289778.1 | 2 - 3 | 568 | 80 | |
| XM_015434293.1 | XP_015289779.1 | 3 - 4 | 576 | 80 | ||
| GenBank mRNA | KP238484.1 | - | 2 - 3 | 446 | 80 | |
| KP238485.1 | - | 2 - 3 | 444 | 80 | ||
| KP238486.1 | - | 2 - 3 | 447 | 80 | ||
| KP238487.1 | - | 2 - 3 | 446 | 80 | ||
More Information
Set Membership: |
|

CDKN1C
Genomic Map