Assay Details
| Gene Symbol: FBXL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554506.2 | XP_005554563.1 | 6 - 7 | 1016 | 69 | |
| XM_015450183.1 | XP_015305669.1 | 6 - 7 | 1016 | 69 | ||
| GenBank mRNA | AB170404.1 | - | 2 - 3 | 240 | 69 | |
More Information
Set Membership: |
|

FBXL5
Genomic Map