Assay Details
| Gene Symbol: SGTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319588.1 | NP_001306517.1 | 1119 | 75 | ||
| XM_005587475.2 | XP_005587532.1 | 7 - 8 | 795 | 75 | ||
| GenBank mRNA | AB169229.1 | - | 1119 | 75 | ||
More Information
Set Membership: |
|

SGTA
Genomic Map