Assay Details
| Gene Symbol: ITLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541233.2 | XP_005541290.1 | 2 - 3 | 258 | 97 | |
| XM_005541234.2 | XP_005541291.1 | 2 - 3 | 241 | 97 | ||
More Information
Set Membership: |
|

ITLN1
Genomic Map