Assay Details
| Gene Symbol: TAF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553245.2 | XP_005553302.2 | 1 - 2 | 293 | 92 | |
| XM_015449541.1 | XP_015305027.1 | 1 - 2 | 1342 | 92 | ||
More Information
Set Membership: |
|

TAF11
Genomic Map