Assay Details
| Gene Symbol: SLC2A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544859.2 | XP_005544916.1 | 2 - 3 | 186 | 81 | |
| XM_005544860.2 | XP_005544917.1 | 2 - 3 | 255 | 81 | ||
More Information
Set Membership: |
|

SLC2A5
Genomic Map