Assay Details
| Gene Symbol: NIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541211.2 | XP_005541268.1 | 2 - 3 | 200 | 71 | |
| XM_005541213.2 | XP_005541270.1 | 2 - 3 | 234 | 71 | ||
| XM_015454641.1 | XP_015310127.1 | 2 - 3 | 554 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005541213.2 transcripts.
Set Membership: |
|

NIT1
Genomic Map