Assay Details
| Gene Symbol: LARP4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564490.2 | XP_005564547.1 | 15 - 16 | 5795 | 58 | |
| XM_005564491.2 | XP_005564548.2 | 17 - 18 | 2783 | 58 | ||
More Information
Set Membership: |
|

LARP4B
Genomic Map