Assay Details
| Gene Symbol: LOC102138874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553648.2 | XP_005553705.1 | 6 - 7 | 1108 | 69 | |
| GenBank mRNA | U02976.1 | - | 6 - 7 | 1014 | 69 | |
More Information
Set Membership: |
|

LOC102138874
Genomic Map