Assay Details
| Gene Symbol: IL18BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015435430.1 | XP_015290916.1 | 2 - 3 | 2247 | 86 | |
| XM_015435431.1 | XP_015290917.1 | 3 - 4 | 1358 | 86 | ||
| XM_015435432.1 | XP_015290918.1 | 4 - 5 | 1470 | 86 | ||
More Information
Set Membership: |
|

IL18BP
Genomic Map