Assay Details
| Gene Symbol: LOC102139510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577486.2 | XP_005577543.2 | 1 - 2 | 221 | 78 | |
| GenBank mRNA | AB072015.1 | - | 1 - 2 | 108 | 78 | |
| AB169142.1 | - | 1 - 2 | 169 | 78 | ||
| AB170272.1 | - | 1 - 2 | 163 | 78 | ||
More Information
Set Membership: |
|

LOC102139510
Genomic Map