Assay Details
| Gene Symbol: NDUFS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587389.2 | XP_005587446.1 | 3 - 4 | 175 | 98 | |
| XM_005587390.2 | XP_005587447.1 | 3 - 4 | 415 | 98 | ||
| XM_015440331.1 | XP_015295817.1 | 3 - 4 | 178 | 98 | ||
More Information
Set Membership: |
|

NDUFS7
Genomic Map