Assay Details
| Gene Symbol: CUTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553301.2 | XP_005553358.1 | 5 - 6 | 615 | 65 | |
| XM_015449558.1 | XP_015305044.1 | 6 - 7 | 515 | 65 | ||
| XM_015449559.1 | XP_015305045.1 | 5 - 6 | 589 | 65 | ||
More Information
Set Membership: |
|

CUTA
Genomic Map