Assay Details
| Gene Symbol: FKBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588463.2 | XP_005588520.1 | 2 - 3 | 398 | 98 | |
| XM_005588464.2 | XP_005588521.1 | 2 - 3 | 398 | 98 | ||
| XM_005588465.2 | XP_005588522.2 | 2 - 3 | 488 | 98 | ||
| XM_015440319.1 | XP_015295805.1 | 2 - 3 | 488 | 98 | ||
More Information
Set Membership: |
|

FKBP8
Genomic Map