Assay Details
| Gene Symbol: ZXDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547905.2 | XP_005547962.1 | 10 - 11 | 2676 | 56 | |
| XM_005547906.2 | XP_005547963.1 | 9 - 10 | 2613 | 56 | ||
More Information
Set Membership: |
|

ZXDC
Genomic Map