Assay Details
| Gene Symbol: TMEM222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283974.1 | NP_001270903.1 | 4 - 5 | 592 | 58 | |
| XM_005544295.2 | XP_005544352.1 | 5 - 6 | 580 | 58 | ||
| GenBank mRNA | AB171318.1 | - | 5 - 6 | 496 | 58 | |
| AB174607.1 | - | 4 - 5 | 592 | 58 | ||
More Information
Set Membership: |
|

TMEM222
Genomic Map