Assay Details
| Gene Symbol: UTP14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594545.2 | XP_005594602.1 | 5 - 6 | 481 | 153 | |
| XM_005594547.2 | XP_005594604.1 | 5 - 6 | 482 | 153 | ||
More Information
Set Membership: |
|

UTP14A
Genomic Map