Assay Details
| Gene Symbol: IFI27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562094.2 | XP_005562151.1 | 4 - 5 | 426 | 122 | |
| XM_005562095.2 | XP_005562152.1 | 4 - 5 | 456 | 122 | ||
| XM_015454168.1 | XP_015309654.1 | 4 - 5 | 394 | 122 | ||
| XM_015454169.1 | XP_015309655.1 | 3 - 4 | 264 | 122 | ||
More Information
Set Membership: |
|

IFI27
Genomic Map