Assay Details
| Gene Symbol: SDHD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579623.2 | XP_005579680.1 | 2 - 3 | 295 | 57 | |
| XM_005579624.1 | XP_005579681.1 | 2 - 3 | 243 | 57 | ||
More Information
Set Membership: |
|

SDHD
Genomic Map