Assay Details
| Gene Symbol: MYCBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543908.2 | XP_005543965.1 | 4 - 5 | 353 | 102 | |
| GenBank mRNA | AB174688.1 | - | 4 - 5 | 318 | 102 | |
More Information
Set Membership: |
|

MYCBP
Genomic Map