Assay Details
| Gene Symbol: HAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005541672.2 | XP_005541729.1 | 3 - 4 | 410 | 158 | |
| XM_005541673.2 | XP_005541730.1 | 2 - 3 | 283 | 158 | ||
More Information
Set Membership: |
|

HAX1
Genomic Map