Assay Details
| Gene Symbol: GRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287635.1 | NP_001274564.1 | 5 - 6 | 792 | 80 | |
| GenBank mRNA | AB170303.1 | - | 5 - 6 | 792 | 80 | |
| AB220412.1 | - | 5 - 6 | 762 | 80 | ||
More Information
Set Membership: |
|

GRB2
Genomic Map