Assay Details
| Gene Symbol: THNSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575346.2 | XP_005575403.1 | 4 - 5 | 927 | 57 | |
| XM_005575347.2 | XP_005575404.1 | 4 - 5 | 670 | 57 | ||
| XM_005575348.2 | XP_005575405.1 | 4 - 5 | 3029 | 57 | ||
| XM_005575349.2 | XP_005575406.1 | 4 - 5 | 670 | 57 | ||
| XM_005575350.2 | XP_005575407.1 | 3 - 4 | 3253 | 57 | ||
| GenBank mRNA | AB171007.1 | - | 4 - 5 | 658 | 57 | |
More Information
Set Membership: |
|

THNSL2
Genomic Map