Assay Details
| Gene Symbol: SLC25A15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585714.2 | XP_005585771.1 | 1 - 2 | 1666 | 107 | |
| XM_005585716.2 | XP_005585773.1 | 1 - 2 | 1668 | 107 | ||
| XM_005585717.2 | XP_005585774.1 | 1 - 2 | 1669 | 107 | ||
More Information
Set Membership: |
|

SLC25A15
Genomic Map