Assay Details
| Gene Symbol: PMM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285089.1 | NP_001272018.1 | 4 - 5 | 415 | 81 | |
| GenBank mRNA | AB169900.1 | - | 4 - 5 | 447 | 81 | |
| AB170337.1 | - | 4 - 5 | 415 | 81 | ||
More Information
Set Membership: |
|

PMM1
Genomic Map