Assay Details
| Gene Symbol: ARL8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005595960.2 | XP_005596017.2 | 7 | 1047 | 141 | |
| GenBank mRNA | AB169820.1 | - | 985 | 141 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARL8B| Gene Symbol: ARL8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005595960.2 | XP_005596017.2 | 7 | 1047 | 141 | |
| GenBank mRNA | AB169820.1 | - | 985 | 141 | ||
Set Membership: |
|