Assay Details
| Gene Symbol: ATP6V0C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015442574.1 | XP_015298060.1 | 2 | 644 | 131 | |
| GenBank mRNA | AB220419.1 | - | 1 | 139 | 131 | |
More Information
Set Membership: |
|

ATP6V0C
Genomic Map