Assay Details
| Gene Symbol: SMARCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584106.2 | XP_005584163.1 | 10 | 2384 | 149 | |
| XM_005584108.2 | XP_005584165.1 | 10 | 2384 | 149 | ||
| XM_005584109.2 | XP_005584166.1 | 8 | 2235 | 149 | ||
More Information
Set Membership: |
|

SMARCE1
Genomic Map