Assay Details
| Gene Symbol: RAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284626.1 | NP_001271555.1 | 4 | 396 | 117 | |
| GenBank mRNA | AB169865.1 | - | 4 | 396 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAN| Gene Symbol: RAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284626.1 | NP_001271555.1 | 4 | 396 | 117 | |
| GenBank mRNA | AB169865.1 | - | 4 | 396 | 117 | |
Set Membership: |
|