Assay Details
| Gene Symbol: MSANTD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581192.2 | XP_005581249.1 | 3 | 979 | 149 | |
| XM_005581193.2 | XP_005581250.1 | 4 | 1371 | 149 | ||
| XM_005581194.2 | XP_005581251.1 | 3 | 1014 | 149 | ||
More Information
Set Membership: |
|

MSANTD3
Genomic Map