Assay Details
| Gene Symbol: DNAJB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588251.2 | XP_005588308.1 | 3 | 1290 | 81 | |
| XM_015440637.1 | XP_015296123.1 | 4 | 1077 | 81 | ||
| XM_015440638.1 | XP_015296124.1 | 4 | 1103 | 81 | ||
| XM_015440639.1 | XP_015296125.1 | 4 | 1165 | 81 | ||
More Information
Set Membership: |
|

DNAJB1
Genomic Map