Assay Details
| Gene Symbol: C1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015430314.1 | XP_015285800.1 | 2 | 293 | 140 | |
| GenBank mRNA | AB169667.1 | - | 2 | 231 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C1R| Gene Symbol: C1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015430314.1 | XP_015285800.1 | 2 | 293 | 140 | |
| GenBank mRNA | AB169667.1 | - | 2 | 231 | 140 | |
Set Membership: |
|