Assay Details
| Gene Symbol: GPX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547106.2 | XP_005547163.2 | 1 | 432 | 150 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005547106.2 transcripts.
Set Membership: |
|

GPX1
Genomic Map