Assay Details
| Gene Symbol: LOC102144128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547321.2 | XP_005547378.1 | 1 - 2 | 3463 | 131 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005547321.2 transcripts.
Set Membership: |
|

LOC102144128
Genomic Map